4,5-Dichloro-6-(trifluoromethyl)pyrimidine
Leverantör: Apollo Scientific
Varning
PC48394-1G PC48394-250MG PC48394-5G
APOSPC48394-1GEA 6775 SEK
APOSPC48394-1G APOSPC48394-250MG APOSPC48394-5G
4,5-Dichloro-6-(trifluoromethyl)pyrimidine
4,5-Dichloro-6-(trifluoromethyl)pyrimidine
Formel: C₅HCl₂F₃N₂
MW: 216,976 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD09909760
CAS nummer: 141602-36-0