4-(Aminomethyl)-2-(trifluoromethyl)pyridine hydrochloride
Leverantör: Apollo Scientific
Varning
PC48382-100MG PC48382-250MG
APOSPC48382-100MGEA 2075 SEK
APOSPC48382-100MG APOSPC48382-250MG
4-(Aminomethyl)-2-(trifluoromethyl)pyridine hydrochloride
4-(Aminomethyl)-2-(trifluoromethyl)pyridine hydrochloride
Formel: C₇H₇F₃N₂·ClH
MW: 212,6 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD12400889
CAS nummer: 1159813-38-3