HFPO oligomer acid fluorides (mixture) 97%
Leverantör: Apollo Scientific
|
Fara
|
PC4819-25G
PC4819-5G
APOSPC4819-25GEA
4125
SEK
APOSPC4819-25G
APOSPC4819-5G
HFPO oligomer acid fluorides (mixture) 97%
HFPO oligomer acid fluorides (mixture)
Storage Temperature:
Omgivande |
MDL Number:
MFCD00163888 CAS nummer: 65208-35-7 UN: 3265 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...