Difluoroacetonitrile 97%
Leverantör: Apollo Scientific
|
Fara
|
Synonyms:
Difluoromethyl cyanide, 2,2-Difluoroethanonitrile
PC4811-25G
PC4811-5G
APOSPC4811-25GEA
8150
SEK
APOSPC4811-25G
APOSPC4811-5G
Difluoroacetonitrile 97%
Difluoroacetonitrile
Formel:
C₂HF₂N MW: 77.033 g/mol Kokpunkt: 23 °C Densitet: 1.113 g/cm³ (25 °C) Storage Temperature: Omgivande |
MDL Number:
MFCD01673031 CAS nummer: 359-12-6 UN: 3275 ADR: 6.1+3 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...