1,2,3,4-Tetrachlorohexafluorocyclopentane 98%
Leverantör: Apollo Scientific
Varning
Synonyms: 1,1,2,3,4,5-Hexafluoro-2,3,4,5-tetrachlorocyclopentane
PC4801-1G PC4801-5G
APOSPC4801-1GEA 1025 SEK
APOSPC4801-1G APOSPC4801-5G
1,2,3,4-Tetrachlorohexafluorocyclopentane 98%
1,2,3,4-Tetrachlorohexafluorocyclopentane
Formel: C₅Cl₄F₆
MW: 315.856 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00042312
CAS nummer: 1453-38-9