2-tert-Butyl-7-piperazino-5-(trifluoromethyl)pyrazolo[1,5-a]pyrimidine 97%
Leverantör: Apollo Scientific
Varning
PC4646-1G PC4646-500MG
APOSPC4646-1GEA 2350 SEK
APOSPC4646-1G APOSPC4646-500MG
2-tert-Butyl-7-piperazino-5-(trifluoromethyl)pyrazolo[1,5-a]pyrimidine 97%
2-tert-Butyl-7-piperazino-5-(trifluoromethyl)pyrazolo[1,5-a]pyrimidine
Formel: C₁₅H₂₀F₃N₅
MW: 327,348 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00728816
CAS nummer: 516494-46-5