2,2-Difluoroethyl hexafluoroisopropyl carbonate
Leverantör: Apollo Scientific
Varning
PC450294-1G PC450294-5G
APOSPC450294-1GEA 1300 SEK
APOSPC450294-1G APOSPC450294-5G
2,2-Difluoroethyl hexafluoroisopropyl carbonate
2,2-Difluoroethyl hexafluoroisopropyl carbonate
Formel: C₆H₄F₈O₃
MW: 276,081 g/mol
Kokpunkt: 80…81 °C (150 mm Hg)
Storage Temperature: Omgivande
MDL Number: MFCD28334289
CAS nummer: 1980044-06-1
UN: 1993
ADR: 3,III