2-(Chlorodifluoromethyl)-4,6-bis(trifluoromethyl)-1,3,5-triazine
Leverantör: Apollo Scientific
PC450253-1G PC450253-25G PC450253-5G
APOSPC450253-1GEA 795 SEK
APOSPC450253-1G APOSPC450253-25G APOSPC450253-5G
2-(Chlorodifluoromethyl)-4,6-bis(trifluoromethyl)-1,3,5-triazine
2-(Chlorodifluoromethyl)-4,6-bis(trifluoromethyl)-1,3,5-triazine
Formel: C₆ClF₈N₃
MW: 301.525 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD27918507
CAS nummer: 328-77-8