Hexakis(1H,1H-trifluoroethoxy)phosphazene
Leverantör: Apollo Scientific
Varning
PC450202-1G PC450202-250MG
APOSPC450202-1GEA 8525 SEK
APOSPC450202-1G APOSPC450202-250MG
Hexakis(1H,1H-trifluoroethoxy)phosphazene
Hexakis(1H,1H-trifluoroethoxy)phosphazene
Formel: C₁₂H₁₂F₁₈N₃O₆P₃
MW: 729,133 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00307380
CAS nummer: 1065-05-0