5-Bromo-1,1,1-trifluoropentane
Leverantör: Apollo Scientific
PC450116-1G
PC450116-5G
APOSPC450116-1GEA
1850
SEK
APOSPC450116-1G
APOSPC450116-5G
5-Bromo-1,1,1-trifluoropentane
5-Bromo-1,1,1-trifluoropentane
Formel:
C₅H₈BrF₃ MW: 205.016 g/mol Storage Temperature: Omgivande |
MDL Number:
MFCD07780194 CAS nummer: 54932-74-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...