5-Bromo-4,4,5,5-tetrafluoropentanamide

Leverantör: Apollo Scientific

PC450097-1G PC450097-5G
APOSPC450097-1GEA 1625 SEK
APOSPC450097-1G APOSPC450097-5G
5-Bromo-4,4,5,5-tetrafluoropentanamide
5-Bromo-4,4,5,5-tetrafluoropentanamide
Formel: C₅H₆BrF₄NO
MW: 252,005 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD18074379
CAS nummer: 1309602-30-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR