Methyl 5-bromo-4,4,5,5-tetrafluoropentanoate

Leverantör: Apollo Scientific
Varning

PC450095-1G PC450095-5G
APOSPC450095-1GEA 1125 SEK
APOSPC450095-1G APOSPC450095-5G
Methyl 5-bromo-4,4,5,5-tetrafluoropentanoate
Methyl 5-bromo-4,4,5,5-tetrafluoropentanoate
Formel: C₆H₇BrF₄O₂
MW: 267,016 g/mol
Kokpunkt: 71…73 °C (8 mm Hg)
Storage Temperature: Omgivande
MDL Number: MFCD18074352
CAS nummer: 1309602-78-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR