2-[1-Methyl-3-(trifluoromethyl)pyrazol-4-yl]-2-oxo-acetic acid 95%
Leverantör: Apollo Scientific
PC44903-1G PC44903-250MG
APOSPC44903-1GEA 10175 SEK
APOSPC44903-1G APOSPC44903-250MG
2-[1-Methyl-3-(trifluoromethyl)pyrazol-4-yl]-2-oxo-acetic acid 95%
2-[1-Methyl-3-(trifluoromethyl)pyrazol-4-yl]-2-oxo-acetic acid
Formel: C₇H₅F₃N₂O₃
MW: 222.121 g/mol
Storage Temperature: Omgivande
CAS nummer: 2141373-00-2