5-Trifluoromethylpyridine-2-carboxamide 95+%
Leverantör: Apollo Scientific
PC446195-5G
PC446195-10G
PC446195-1G
APOSPC446195-5GEA
6100
SEK
APOSPC446195-5G
APOSPC446195-10G
APOSPC446195-1G
5-Trifluoromethylpyridine-2-carboxamide 95+%
5-Trifluoromethylpyridine-2-carboxamide
Formel:
C₇H₅F₃N₂O MW: 190,123 g/mol Storage Temperature: Omgivande |
MDL Number:
MFCD02656019 CAS nummer: 22245-86-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...