4-(Trifluoromethyl)pyridine-2-carboxylic acid methyl ester 95+%
Leverantör: Apollo Scientific
Varning
Synonyms: Methyl 4-(trifluoromethyl)picolinate
PC446183-1G PC446183-250MG PC446183-5G
APOSPC446183-1GEA 2875 SEK
APOSPC446183-1G APOSPC446183-250MG APOSPC446183-5G
4-(Trifluoromethyl)pyridine-2-carboxylic acid methyl ester 95+%
4-(Trifluoromethyl)pyridine-2-carboxylic acid methyl ester
Formel: C₈H₆F₃NO₂
Storage Temperature: Omgivande
MDL Number: MFCD11113409
CAS nummer: 455941-78-3