3-Fluoroisonicotinic acid tert-butyl ester 95+%

Leverantör: Apollo Scientific

PC446139-1G
APOSPC446139-1GEA 9725 SEK
APOSPC446139-1G
3-Fluoroisonicotinic acid tert-butyl ester 95+%
3-Fluoroisonicotinic acid tert-butyl ester
Formel: C₁₀H₁₂FNO₂
MW: 197.206 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD21114998
CAS nummer: 1254475-68-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR