2-Fluoroisonicotinic acid tert-butyl ester 95+%

Leverantör: Apollo Scientific

PC446138-1G
APOSPC446138-1GEA 7050 SEK
APOSPC446138-1G
2-Fluoroisonicotinic acid tert-butyl ester 95+%
2-Fluoroisonicotinic acid tert-butyl ester
Formel: C₁₀H₁₂FNO₂
MW: 197.206 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD18897596
CAS nummer: 1339544-40-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR