3-(6-Methyl-4-trifluoromethyl-pyridin-2-yl)-benzoic acid methyl ester
Leverantör: Apollo Scientific
PC446056-1G PC446056-500MG
APOSPC446056-1GEA 4550 SEK
APOSPC446056-1G APOSPC446056-500MG
3-(6-Methyl-4-trifluoromethyl-pyridin-2-yl)-benzoic acid methyl ester
3-(6-Methyl-4-trifluoromethyl-pyridin-2-yl)-benzoic acid methyl ester
Formel: C₁₅H₁₂F₃NO₂
MW: 295.256 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD19981343
CAS nummer: 1311280-43-9