N-Methyl-N-(6-morpholin-4'-yl-3-nitro-4-(trifluoromethyl)pyridin-2-yl)hydrazine
Leverantör: Apollo Scientific
PC446029-1G PC446029-5G
APOSPC446029-1GEA 3625 SEK
APOSPC446029-1G APOSPC446029-5G
N-Methyl-N-(6-morpholin-4'-yl-3-nitro-4-(trifluoromethyl)pyridin-2-yl)hydrazine
N-Methyl-N-(6-morpholin-4'-yl-3-nitro-4-(trifluoromethyl)pyridin-2-yl)hydrazine
Formel: C₁₁H₁₄F₃N₅O₃
MW: 321.256 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD11052406
CAS nummer: 1053658-19-7