3,5-Difluoro-4-iodonitrobenzene
Leverantör: Apollo Scientific
Varning
Synonyms: 1,3-Difluoro-2-iodo-5-nitrobenzene
PC4246-1G PC4246-250MG
APOSPC4246-1GEA 4125 SEK
APOSPC4246-1G APOSPC4246-250MG
3,5-Difluoro-4-iodonitrobenzene
3,5-Difluoro-4-iodonitrobenzene
Formel: C₆H₂F₂INO₂
MW: 284,987 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD07368756
CAS nummer: 886762-63-6