Ethyl 2-amino-4,5-difluorobenzoate 97%
Leverantör: Apollo Scientific
PC421229-10G PC421229-1G PC421229-5G
APOSPC421229-10GEA 15025 SEK
APOSPC421229-10G APOSPC421229-1G APOSPC421229-5G
Ethyl 2-amino-4,5-difluorobenzoate 97%
Ethyl 2-amino-4,5-difluorobenzoate
Formel: C₉H₉F₂NO₂
MW: 201,17 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD11226323
CAS nummer: 864293-36-7