(1R,2R)-2-(3,4-Difluorophenyl)cyclopropanecarboxamide 98%
Leverantör: Apollo Scientific
PC421186-100MG PC421186-250MG
APOSPC421186-100MGEA 8250 SEK
APOSPC421186-100MG APOSPC421186-250MG
(1R,2R)-2-(3,4-Difluorophenyl)cyclopropanecarboxamide 98%
(1R,2R)-2-(3,4-Difluorophenyl)cyclopropanecarboxamide
Formel: C₁₀H₉F₂NO
MW: 197.181 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD22380631
CAS nummer: 1006376-62-0