2-Chloro-5-(difluoromethoxy)pyridine
Leverantör: Apollo Scientific
Varning
PC421027-1G PC421027-500MG
APOSPC421027-1GEA 1700 SEK
APOSPC421027-1G APOSPC421027-500MG
2-Chloro-5-(difluoromethoxy)pyridine
2-Chloro-5-(difluoromethoxy)pyridine
Formel: C₆H₄ClF₂NO
MW: 179.552 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD13185849
CAS nummer: 1206980-28-0