(6S)-1,1-Difluoro-5-azaspiro[2.4]heptane-6-carboxylic acid, N-BOC protected
Leverantör: Apollo Scientific
Varning
PC409047-1G PC409047-100MG PC409047-250MG
APOSPC409047-1GEA 15575 SEK
APOSPC409047-1G APOSPC409047-100MG APOSPC409047-250MG
(6S)-1,1-Difluoro-5-azaspiro[2.4]heptane-6-carboxylic acid, N-BOC protected
(6S)-1,1-Difluoro-5-azaspiro[2.4]heptane-6-carboxylic acid, N-BOC protected
Formel: C₁₂H₁₇F₂NO₄
MW: 277,264 g/mol
Smältpunkt: 87…97 °C
Storage Temperature: Omgivande
MDL Number: MFCD21362512
CAS nummer: 1357482-03-1