6-Chloro-5-fluoro-2-methylpyrimidin-4-amine
Leverantör: Apollo Scientific
PC408560-1G PC408560-250MG
APOSPC408560-1GEA 960 SEK
APOSPC408560-1G APOSPC408560-250MG
6-Chloro-5-fluoro-2-methylpyrimidin-4-amine
6-Chloro-5-fluoro-2-methylpyrimidin-4-amine
Formel: C₅H₅ClFN₃
MW: 161.565 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD11846146
CAS nummer: 943006-45-9