4,6-Difluoro-5-methylpyrimidine
Leverantör: Apollo Scientific
PC408556-1G PC408556-250MG
APOSPC408556-1GEA 1875 SEK
APOSPC408556-1G APOSPC408556-250MG
4,6-Difluoro-5-methylpyrimidine
4,6-Difluoro-5-methylpyrimidine
Formel: C₅H₄F₂N₂
MW: 130,095 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD11656645
CAS nummer: 18260-64-5