2-Bromo-3,4-difluorobenzoic acid
Leverantör: Apollo Scientific
Varning
PC408284-1G PC408284-5G PC408284-25G
APOSPC408284-1GEA 175 SEK
APOSPC408284-1G APOSPC408284-5G APOSPC408284-25G
2-Bromo-3,4-difluorobenzoic acid
2-Bromo-3,4-difluorobenzoic acid
Formel: C₇H₃BrF₂O₂
MW: 236,998 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD09835210
CAS nummer: 170108-05-1
UN: 3077
ADR: 9+EHS,III