4,4,5,5,5-Pentafluoropent-2-en-1-ol 94%

Leverantör: Apollo Scientific
Varning

PC408155-25G PC408155-5G
APOSPC408155-25GEA 10650 SEK
APOSPC408155-25G APOSPC408155-5G
4,4,5,5,5-Pentafluoropent-2-en-1-ol 94%
4,4,5,5,5-Pentafluoropent-2-en-1-ol
Formel: C₅H₅F₅O
MW: 176,085 g/mol
Kokpunkt: 134 °C
Storage Temperature: Omgivande
MDL Number: MFCD24842572
CAS nummer: 148757-93-1
UN: 1987
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR