2-(2-Fluoro-4-iodoanilino)-3,4-difluorobenzoic acid 98%
Leverantör: Apollo Scientific
Varning
Synonyms: 3,4-Difluoro-2-((2-fluoro-4-iodophenyl)amino)benzoic acid
PC408065-250MG PC408065-1G PC408065-5G
APOSPC408065-250MGEA 1250 SEK
APOSPC408065-250MG APOSPC408065-1G APOSPC408065-5G
2-(2-Fluoro-4-iodoanilino)-3,4-difluorobenzoic acid 98%
2-(2-Fluoro-4-iodoanilino)-3,4-difluorobenzoic acid
Formel: C₁₃H₇F₃INO₂
MW: 393,1 g/mol
Smältpunkt: 195…200 °C
Storage Temperature: Kylskåp
MDL Number: MFCD08690072
CAS nummer: 391211-97-5