(3-Fluorobicyclo[1.1.1]pentan-1-yl)methanol 97%
Leverantör: Apollo Scientific
Varning
Synonyms: {3-fluorobicyclo[1.1.1]pentan-1-yl}methanol
PC405698-250MG PC405698-100MG
APOSPC405698-250MGEA 29925 SEK
APOSPC405698-250MG APOSPC405698-100MG
(3-Fluorobicyclo[1.1.1]pentan-1-yl)methanol 97%
(3-Fluorobicyclo[1.1.1]pentan-1-yl)methanol
Formel: C₆H₉FO
MW: 116.133 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD29037345
CAS nummer: 262852-01-7