3-([Pentafluorothio)benzeneboronic acid, pinacol ester
Leverantör: Apollo Scientific
Varning
Synonyms: 4,4,5,5-Tetramethyl-2-(m-pentafluorosulphanylbenzene)-1,3,2-dioxaborolane
PC405653-1G PC405653-250MG PC405653-5G
APOSPC405653-1GEA 8475 SEK
APOSPC405653-1G APOSPC405653-250MG APOSPC405653-5G
3-([Pentafluorothio)benzeneboronic acid, pinacol ester
3-([Pentafluorothio)benzeneboronic acid, pinacol ester
Formel: C₁₂H₁₆BF₅O₂S
MW: 330,122 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD20489193
CAS nummer: 1286278-35-0