6-Fluoro-1,2,4-triazolo[4,3-a]pyrimidine-5,7(1H,6H)-dione
Leverantör: Apollo Scientific
PC404015-1G
PC404015-250MG
APOSPC404015-1GEA
5075
SEK
APOSPC404015-1G
APOSPC404015-250MG
6-Fluoro-1,2,4-triazolo[4,3-a]pyrimidine-5,7(1H,6H)-dione
6-Fluoro-1,2,4-triazolo[4,3-a]pyrimidine-5,7(1H,6H)-dione
Formel:
C₅H₃FN₄O₂ MW: 170,101 g/mol Storage Temperature: Omgivande |
MDL Number:
MFCD32264095 CAS nummer: 105441-05-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...