3-[2-(Trifluoromethyl)phenyl]oxetan-3-amine hydrochloride 97%
Leverantör: Apollo Scientific
PC403037-1G PC403037-250MG PC403037-500MG
APOSPC403037-1GEA 11375 SEK
APOSPC403037-1G APOSPC403037-250MG APOSPC403037-500MG
3-[2-(Trifluoromethyl)phenyl]oxetan-3-amine hydrochloride 97%
3-[2-(Trifluoromethyl)phenyl]oxetan-3-amine hydrochloride
Formel: C₁₀H₁₀F₃NO·ClH
MW: 253,649 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD20233042
CAS nummer: 1349715-87-2