3-Chloro-1-methyl-5-(trifluoromethyl)pyridin-2-one 95%
Leverantör: Apollo Scientific
Varning
PC401046-1G PC401046-250MG PC401046-5G
APOSPC401046-1GEA 5275 SEK
APOSPC401046-1G APOSPC401046-250MG APOSPC401046-5G
3-Chloro-1-methyl-5-(trifluoromethyl)pyridin-2-one 95%
3-Chloro-1-methyl-5-(trifluoromethyl)pyridin-2-one
Formel: C₇H₅ClF₃NO
MW: 211.569 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD28122856
CAS nummer: 1281189-85-2