1-(Tetrahydrofuran-3-yl)piperazine bis(2,2,2-trifluoroacetate)
Leverantör: Apollo Scientific
Varning
PC400592-1G PC400592-500MG
APOSPC400592-1GEA 18175 SEK
APOSPC400592-1G APOSPC400592-500MG
1-(Tetrahydrofuran-3-yl)piperazine bis(2,2,2-trifluoroacetate)
1-(Tetrahydrofuran-3-yl)piperazine bis(2,2,2-trifluoroacetate)
Formel: C₈H₁₆N₂O·2CF₃CO₂H
MW: 384.272 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD23106360
CAS nummer: 1403767-15-6