(1R,2S)-2-Fluorocyclopropanecarboxylic acid 97%
Leverantör: Apollo Scientific
Fara
PC400539-100MG PC400539-1G PC400539-250MG
APOSPC400539-100MGEA 3775 SEK
APOSPC400539-100MG APOSPC400539-1G APOSPC400539-250MG
(1R,2S)-2-Fluorocyclopropanecarboxylic acid 97%
(1R,2S)-2-Fluorocyclopropanecarboxylic acid
Formel: C₄H₅FO₂
MW: 104,08 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD19237658
CAS nummer: 167073-08-7
UN: 3265
ADR: 8,II