(2-Chloro-5-(trifluoromethyl)pyridin-3-yl)methanol
Leverantör: Apollo Scientific
Varning
PC400187-1G PC400187-250MG
APOSPC400187-1GEA 2975 SEK
APOSPC400187-1G APOSPC400187-250MG
(2-Chloro-5-(trifluoromethyl)pyridin-3-yl)methanol
(2-Chloro-5-(trifluoromethyl)pyridin-3-yl)methanol
Formel: C₇H₅ClF₃NO
MW: 211.569 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD11848356
CAS nummer: 943551-28-8