8-Bromo-2-(trifluoromethyl)imidazo[1,2-a]pyridine-3-carboxylic acid
Leverantör: Apollo Scientific
Varning
PC37856-1G PC37856-250MG
APOSPC37856-1GEA 7175 SEK
APOSPC37856-1G APOSPC37856-250MG
8-Bromo-2-(trifluoromethyl)imidazo[1,2-a]pyridine-3-carboxylic acid
8-Bromo-2-(trifluoromethyl)imidazo[1,2-a]pyridine-3-carboxylic acid
Formel: C₉H₄BrF₃N₂O₂
MW: 309.039 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD18073566
CAS nummer: 1038389-93-3