1-(2-Fluorophenyl)-imidazolidin-2-one ≥95%
Leverantör: Apollo Scientific
Varning
PC32976-1G PC32976-250MG
APOSPC32976-1GEA 4550 SEK
APOSPC32976-1G APOSPC32976-250MG
1-(2-Fluorophenyl)-imidazolidin-2-one ≥95%
1-(2-Fluorophenyl)-imidazolidin-2-one
Formel: C₉H₉FN₂O
MW: 180,179 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD23696665
CAS nummer: 1190281-65-2