4-(3,4,5-Trifluorophenyl)-1H-1,2,4-triazol-5(4H)-one
Leverantör: Apollo Scientific
Varning
PC32899-1G PC32899-250MG
APOSPC32899-1GEA 990 SEK
APOSPC32899-1G APOSPC32899-250MG
4-(3,4,5-Trifluorophenyl)-1H-1,2,4-triazol-5(4H)-one
4-(3,4,5-Trifluorophenyl)-1H-1,2,4-triazol-5(4H)-one
Formel: C₈H₄F₃N₃O
MW: 215.132 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD31742301
CAS nummer: 2379918-34-8