5-(Trifluoromethyl)-2,3-dihydropyrrolo[2,3-b]pyridine, N1-BOC protected
Leverantör: Apollo Scientific
Varning
PC32795-1G PC32795-250MG
APOSPC32795-1GEA 9025 SEK
APOSPC32795-1G APOSPC32795-250MG
5-(Trifluoromethyl)-2,3-dihydropyrrolo[2,3-b]pyridine, N1-BOC protected
5-(Trifluoromethyl)-2,3-dihydropyrrolo[2,3-b]pyridine, N1-BOC protected
Formel: C₁₃H₁₅F₃N₂O₂
MW: 288.266 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD31812100
CAS nummer: 2379918-60-0