4-Fluoro-3-cyclopropylphenol
Leverantör: Apollo Scientific
Varning
PC32778-1G PC32778-250MG
APOSPC32778-1GEA 820 SEK
APOSPC32778-1G APOSPC32778-250MG
4-Fluoro-3-cyclopropylphenol
4-Fluoro-3-cyclopropylphenol
Formel: C₉H₉FO
MW: 152.166 g/mol
Storage Temperature: Frys
MDL Number: MFCD31700582
CAS nummer: 1887175-94-1