Cyclopentyliden(dimethyl)ammonium tetrafluoroborate

Leverantör: Apollo Scientific

PC31730-1G PC31730-250MG
APOSPC31730-1GEA 1175 SEK
APOSPC31730-1G APOSPC31730-250MG
Cyclopentyliden(dimethyl)ammonium tetrafluoroborate
Cyclopentyliden(dimethyl)ammonium tetrafluoroborate
Formel: C₇H₁₄N·BF₄
MW: 198,997 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00219098
CAS nummer: 95193-30-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR