Ethyl 5-cyano-6-methyl-2-(trifluoromethyl)nicotinate
Leverantör: Apollo Scientific
Fara
PC31134-1G PC31134-250MG
APOSPC31134-1GEA 2200 SEK
APOSPC31134-1G APOSPC31134-250MG
Ethyl 5-cyano-6-methyl-2-(trifluoromethyl)nicotinate
Ethyl 5-cyano-6-methyl-2-(trifluoromethyl)nicotinate
Formel: C₁₁H₉F₃N₂O₂
MW: 258.197 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD01312037
CAS nummer: 445-71-6
UN: 2811
ADR: 6.1 III