5-Bromo-2-methyl-3-(trifluoromethyl)benzoic acid 97%
Leverantör: Apollo Scientific
PC303592-1G PC303592-5G
APOSPC303592-1GEA 3625 SEK
APOSPC303592-1G APOSPC303592-5G
5-Bromo-2-methyl-3-(trifluoromethyl)benzoic acid 97%
5-Bromo-2-methyl-3-(trifluoromethyl)benzoic acid
Formel: C₉H₆BrF₃O₂
MW: 283.042 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD27942784
CAS nummer: 1266728-50-0