2-Bromo-4,6-bis(trifluoromethyl)phenylacetic acid
Leverantör: Apollo Scientific
PC303532-1G PC303532-5G
APOSPC303532-1GEA 0 SEK
APOSPC303532-1G APOSPC303532-5G
2-Bromo-4,6-bis(trifluoromethyl)phenylacetic acid
2-Bromo-4,6-bis(trifluoromethyl)phenylacetic acid
Formel: C₁₀H₅BrF₆O₂
MW: 351.04 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD28728131
CAS nummer: 1807217-12-4