6-Bromo-2,3-difluorobenzamide
Leverantör: Apollo Scientific
PC303437-1G PC303437-5G
APOSPC303437-1GEA 3100 SEK
APOSPC303437-1G APOSPC303437-5G
6-Bromo-2,3-difluorobenzamide
6-Bromo-2,3-difluorobenzamide
Formel: C₇H₄BrF₂NO
MW: 236,014 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD28099964
CAS nummer: 1502530-72-4