2-Fluoro-3-methyl-5-(trifluoromethyl)benzoic acid 97%
Leverantör: Apollo Scientific
PC303374-1G PC303374-5G
APOSPC303374-1GEA 4100 SEK
APOSPC303374-1G APOSPC303374-5G
2-Fluoro-3-methyl-5-(trifluoromethyl)benzoic acid 97%
2-Fluoro-3-methyl-5-(trifluoromethyl)benzoic acid
Formel: C₉H₆F₄O₂
MW: 222.136 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD31916391
CAS nummer: 2387220-05-3