4-Chloro-2-fluoro-5-methylaniline 97%
Leverantör: Apollo Scientific
PC303328-5G PC303328-1G
APOSPC303328-5GEA 1725 SEK
APOSPC303328-5G APOSPC303328-1G
4-Chloro-2-fluoro-5-methylaniline 97%
4-Chloro-2-fluoro-5-methylaniline
Formel: C₇H₇ClFN
MW: 159,589 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD09062485
CAS nummer: 116759-33-2