3,6-Difluoro-2-methoxybenzyl bromide 97%
Leverantör: Apollo Scientific
PC303315-5G PC303315-1G
APOSPC303315-5GEA 8100 SEK
APOSPC303315-5G APOSPC303315-1G
3,6-Difluoro-2-methoxybenzyl bromide 97%
3,6-Difluoro-2-methoxybenzyl bromide
Formel: C₈H₇BrF₂O
MW: 237.041 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD18394899
CAS nummer: 1261645-28-6